Imprimer
Chemical preservative glass ampoules
Fournisseur: EP Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
EPSCACS-2EA 125 EUR
EPSCACS-2
Chemical preservative glass ampoules
Save the time and tedium associated with diluting and aliquoting chemicals for environmental sample preservation by using pre-mixed chemical preservative glass ampoules.
  • Glass ampoule containing a pre-measured aliquot of preservative chemical solution
  • Ampoules are machine-welded closed after filling for a consistent, leak-tight seal
Our pre-formulated and aliquoted glass ampoules are ready for field use. Just break the ampoule at the pre-scored neck area using the provided 'breaker' tool, and pour into the designated sample bottle. Choose from our standard offering or let us custom-fill ampoules according to your personal analytical needs.
Certifications: Manufactured under an ISO certified quality management system.
Cdt: Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification.