Imprimer
1-Step™, Pierce™ TMB-Blotting substrate solution
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PIER34018EA 191 EUR
PIER34018
1-Step™, Pierce™ TMB-Blotting substrate solution
Réactifs d'électrophorèse Western Blotting Reagents
The 1-Step TMB-Blotting is a one component horseradish peroxidase (HRP) substrate for Western blotting and immunohistochemistry. TMB is highly sensitive and generates a blue-purple precipitate. This substrate is provided ready to use for maximum convenience.