Imprimer
Atomic absorption spectrocopy (AAS) standards, sodium in oil, Certipur®, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Certipur®
1.15058.0100EA 315 EUR
1.15058.0100
Atomic absorption spectrocopy (AAS) standards, sodium in oil, Certipur®, Supelco®
Étalons de référence Étalons cationiques Standards de sodium
Standard dissolved in oil is ready-to-use calibration standards for the analysis of sodium by AAS in a non-aqueous matrix.