Imprimer
Atomic absorption spectrocopy (AAS) standards, sodium in oil, Certipur®, Supelco®
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.15058.0100EA 395 EUR
MERC1.15058.0100
Atomic absorption spectrocopy (AAS) standards, sodium in oil, Certipur®, Supelco®
Standard dissolved in oil is ready-to-use calibration standards for the analysis of sodium by AAS in a non-aqueous matrix.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022