Imprimer
AMREE540
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E540-1LEA 80.1 EUR
VWRCE540-1L
AMREE540
Contains 1,5 M NaCl and 0,5 M NaOH. Use to treat Southern blots to yield single-stranded DNA molecules and facilitate probe hybridization.