Imprimer
HPLC columns, Ascentis® RP-Amide, Supelco®, 3 µm
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Ascentis®
565320-UEA 564 EUR
565320-U 565322-U 565321-U 565300-U 565301-U 565313-U 565302-U 565312-U
HPLC columns, Ascentis® RP-Amide, Supelco®, 3 µm
Colonnes pour chromatographie
Ascentis® RP-Amide is a ultra low-bleed, embedded polar group (EPG) phase that provides orthogonal selectivity and increased resolution for polar compounds. The unique selectivity is due to the hydrogen bonding and polar interactions of the stationary phase