Imprimer
Tablettes vidéos, Moticam® BTW
Fournisseur: Motic
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MOTI1100600100811EA 635 EUR
MOTI1100600100811
Tablettes vidéos, Moticam® BTW
The Moticam BTW is a tablet solution with a adjustable bracket to mount your own tablet.
  • Help microscope users to visualize, edit and share the microscope images
  • USB (Windows) or Wi-Fi (iOS or Android) connection
  • C-mount lens, progressive scanning mode
Informations de commande: Tablet is not included. Users can use their own tablet to connect through Wi-Fi.
Informations de livraison: Supplied with USB cable, Image Plus 3.0 software, calibration slide and power supply.