Imprimer

Picro-Sirius Red solution stain for histology

Marques : KLINIPATH (PL)
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

VWR®
VWRK640745EA 39 EUR
VWRK640745
Picro-Sirius Red solution stain for histology
Direct red 80

Sirius red 0,1% in saturated picric acid.


Suitable for use in histology.

Formule: C₄₅H₂₆N₁₀Na₆O₂₁S₆
Numéro MDL: MFCD00054389
Numéro CAS: 2610-10-8
IVDLogo

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR