Imprimer
Screw thread vial convenience kits
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NSCAC5000-95WEA 97.6 EUR
NSCAC5000-95W NSCAC5000-80W NSCAC5000-92W NSCAC5000-86W
Screw thread vial convenience kits
Fioles
These autosampler vial kits are available with assembled or unassembled closures.
  • Save time during sample preparation and reduce the risk of contamination
Reusable two-compartment trays protect vials and closures while keeping matching supplies together. Clear trays to make it easy to keep track of available supplies without opening containers.
Informations de livraison: Each kit includes 100 glass vials and 100 screw thread caps with pre-assembled septa.
Cdt: The kit is packaged in convenient vial trays with clear covers or in economical polybags.