Imprimer
Dowsil™ / Dow Corning 340 Heat Sink Compound
Marques : DOW CORNING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
632272MEA 58.3 EUR
632272M
Dowsil™ / Dow Corning 340 Heat Sink Compound
Dowsil™ / Dow Corning 340 Heat Sink Compound
Dowsil™ / Dow Corning 340 Heat Sink Compound is suitable for thermal coupling of electrical/electronic devices to heat sinks.
  • Non-flowing
  • Moderate thermal conductivity
  • No need for ovens or curing
  • Heat flow away from electronic components can increase reliability
  • Automated or manual dispensing
White, non curing and non flowing thermally conductive compound with composition of Zinc oxide and polydimethylsiloxane
Densité: 2,1
UN: 3077
ADR: ,