Imprimer
Vector, pUC 18
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ICNA04821226EA 204 EUR
ICNA04821226
Vector, pUC 18
Vecteurs, plasmides et librairies
pUC 18 is a small vector of approximately 2,7 Kb containing the Pvu II/EcoR I fragment of pBR322 and the lacZ gene multiple cloning site from M13mp18.