Imprimer
Bleu trypan, vert foncé, poudre vert foncé
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ICNA0219553280EA 132 EUR
ICNA0219553280 ICNA0219553205
Bleu trypan, vert foncé, poudre vert foncé
Bleu trypan
Trypan blue is a blue acid dye with a strong affinity for cellulose containing substrates such as cotton; less affinity for proteinaceous materials.
Trypan blue is used as a vital dye which is especially important because it is taken up by the reticuloendothelial system.Trypan blue is also recommended for use in dye exclusion procedures for viable cell counting. Non-viable cells will up-take trypan blue at a faster rate than viable cells.
Soluble in ethylene glycol monomethyl ether (methyl cellosolve; EGME) (20 mg/ml); Slightly soluble in water or ethanol (0,6 mg/ml); almost insoluble in alcohol.
Formule: C₃₄H₂₄N₆Na₄O₁₄S₄
Poids moléculaire: 960,82 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00003969
Numéro CAS: 72-57-1