Imprimer
Anti-IgG PAN Mouse Monoclonal Antibody [clone: HP6017]
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ICNA08630722EA 617 EUR
ICNA08630722
Anti-IgG PAN Mouse Monoclonal Antibody [clone: HP6017]
Anticorps
This purified monoclonal antibody is supplied in 10 mM phosphate buffered saline, pH 7,4.
Type: Secondary
Antigen: IgG Fc PAN
Clonality: Monoclonal
Clone: HP6017
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2ak
Reactivity: Human