Imprimer
Safety blood lancets, VITREX® STERILANCE® Flex
Fournisseur: Vitrex Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MODU399813EA 338 EUR
MODU399813
Safety blood lancets, VITREX® STERILANCE® Flex
Safety lancets for blood glucose and other in vitro diagnostic tests.
  • Needle fully shielded before and after use
  • Pre-loaded pressure activated design
  • High quality, ultra sharp tri-bevel needle, high speed penetration
Configurations and penetration depths suit most capillary blood requirements.