Imprimer
DOWC634841J
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DOWC634841JEA 1540 EUR
DOWC634841J
DOWC634841J
SILASTIC M RTV Silicone Rubber is a two-component material consisting of a SILASTIC M RTV Base, which when mixed with the SILASTIC M RTV Curing Agent cures at room temperature by an addition reaction
High durometer mold making silicone rubber suited for the reproduction of architectural, furniture components and prototypes.
Attention: Chemically equivalent to XIAMETER® RTV-4136-M base and curing agent.