Imprimer
Pallets and sumps for IBC station
Fournisseur: New Pig
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NEWPPAKE235SW-BKEA 972 EUR
NEWPPAKE235SW-BK
Pallets and sumps for IBC station
The moulded-in sump catches leaks, drips and spills to helps to comply with regulations and keeps the storage areas cleanand safe. Two way forklift entry allows easy movement from either direction (unloaded only).
  • Lift out grates for easy sump access and inspection
  • Central support under grate to maintain UDL capacity
  • Compact footprint reduces floor space needed for IBC storage