Imprimer
Beakers, heavy duty, Berzelius, tall form, borosilicate glass, PYREX®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BIBB1004-600EA 140 EUR
BIBB1004-600 BIBB1004-250 BIBB1004-150
Beakers, heavy duty, Berzelius, tall form, borosilicate glass, PYREX®
Béchers
Made of PYREX® borosilicate glass.
  • Thicker walls and heavier bases
  • Reinforced rims and spout, less prone to breakage during washing
  • Printed with Trace Code for downloadable batch certificate
These heavy duty beakers with white graduations and marking spot are designed for longer life and increased user safety.