Hygromycine B, Millipore®
Marques : Merck Millipore CalbiochemTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Hygromycin B is an aminoglycoside antibiotic that inhibits the growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis by interfering with the translocation of the 70S ribosome and causes misreading of mRNA.
- Cell culture tested
- Sterile-filtered solution in PBS
Formule:
C₂₀H₃₇N₃O₁₃ Poids moléculaire: 527,5 g/mol |
Numéro MDL:
MFCD06795479 Numéro CAS: 31282-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...