Imprimer
Amide resin, NovaPEG Rink
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Novabiochem®
01-64-0473-1EA 106 EUR
01-64-0473-1
Amide resin, NovaPEG Rink
Sorbants pour chromatographie Sorbants SPE
An acid-labile resin for the continuous flow and batch Fmoc SPPS of peptide amides. This resin is prepared by derivatisation of NovaPEG amino resin with the modified Rink linker. The support has excellent swelling properties in a wide range of solvents, including water, MeCN, MeOH, DCM, DMF, THF, and toluene.
  • Has a bead like appearance
  • Appearance ranges from white to light yellow to beige
The high hydrophilicity was also found to benefit on-resin immunoassays with one-bead-one-peptide libraries. Can be stored at +2 to +8 °C.