Imprimer
Ice cubers, self contained
Fournisseur: Hoshizaki
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HOSHIM-65NEEA 3965 EUR
HOSHIM-65NE
Ice cubers, self contained
Machines à glace
The IM-65NE is a self contained ice cube maker is a manual water fill. This Hoshizaki ice makers are micro computer controlled, controlling the ice making process to perform it under varying circumstances, without having to make physical adjustments.
  • A closed water circuit prevent contamination during ice making process
  • A stainless steel exterior with integrated door handles provides a hygienic and high quality finish
  • Foam injected polyurethane for insulation provides the quality and consistency of the ice
  • Removable door gaskets prevent heat and contamination from entering the bin