Imprimer
Protein immunodetection reagents, SuperSignal® West Pico chemiluminescent substrate
Fournisseur: Thermo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PIER34078EA 371 EUR
PIER34078 PIER34079 PIER34087
Protein immunodetection reagents, SuperSignal® West Pico chemiluminescent substrate
SuperSignal® West Pico chemiluminescent substrates use an enhanced chemiluminescence to obtain exceptional sensitivity and signal duration.
  • Highly sensitive for the rapid development of a wide range of protein levels
  • Long signal duration allows time for optimisation of exposure conditions, often negating the need to rerun samples and repeat the blotting procedure
  • Working solution stable for at least 24 hours and the kit can be stored at room temperature, saving valuable cold room space
  • Primary and secondary antibodies are diluted further so they can be used for more blots