Imprimer
Background eliminator, Erase-It®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PIER21065EA 212 EUR
PIER21065
Background eliminator, Erase-It®
Matériel pour le traitement des films
High background, shading, overexposed bands and speckling are problems inherent to film exposure. The Erase-It® kit can correct these problems without the need to re-expose the blot to film or re-do the experiment. Erase-It® solution can be used with newly exposed films or exposed films that have been stored. It can also be used with any brand of film.
  • Fast, easy background elimination from film
  • Eliminates unwanted background from overexposed, speckled or shaded films
  • No need for time-consuming re-exposures to find the optimal image
  • No need to re-optimise assay reagents in order to obtain the optimal image