Imprimer
Chemical resistant cape hood, model 508, MICROCHEM® 3000
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Microchem® 3000
ANSEYE30W-00508-00EA 506 EUR
ANSEYE30W-00508-00
Chemical resistant cape hood, model 508, MICROCHEM® 3000
Vêtements à usage unique
This Balaclava style cape hood covering part of shoulders is made from an external film barrier layer with two non woven layers underneath. The fabric provides a strong liquid and particle barrier.
  • PVC visor
  • Elasticated face opening
  • Ultrasonically welded seams
  • Multi-layer barrier fabric is effective against numerous chemicals
  • Highly visible: Bright yellow for improved worker safety
  • Lightweight yet durable
Typical applications include: Chemicals, oil and petrochemicals, pharmaceutical, food industry (caustic clean downs), sewage purification installations, industrial and tank cleaning, mining.
Certifications: Type 3-B/4-B/5-B, EN 14126, EN 1073-2, EN 1149-5