Imprimer
Homogeniser, dounce type
Fournisseur: BioVision
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BIOV1998-1EA 355 EUR
BIOV1998-1
Homogeniser, dounce type
The tissue grinder is used for homogenising delicate tissues such as intestines, liver, heart and cells.
  • Designed for hand operation
  • Hollow handle permits packing with ice
  • Features expanded reservoir and pouring lip
  • Manually operated, interchangeable pestles
Tubes are made of low extractable borosilicate glass. Clearance between pestle and grinding chamber is 0,09 to 0,16 mm.
Certifications: Glass meets ASTM type 1 Class A and USP Type 1 standards.