Imprimer
General purpose gloves, Monysoft 1
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MARGMFT1100000EA 0 EUR
MARGMFT1100000
General purpose gloves, Monysoft 1
Gants
Glove knitted with a seamless non-stop process. Textile liner made of polyamide (nylon) thread with a high elasticity.
  • Elastic knitwrist
  • Ambidextrous shape doubles the life of the glove by enabling the use of both sides
  • Excellent adaptability owing to its extensibility
  • Low linting
  • Excellent comfort and dexterity
Typical applications include: Delicate manipulation, fine assembly of dry parts, general handling, packaging.
Certifications: Food contact approved