Imprimer
Industrial gloves, K2000BR
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MARGM9999257EA 94.6 EUR
MARGM9999257 MARGM9999254 MARGM9999255 MARGM9999256
Industrial gloves, K2000BR
Gants
Seamless polycotton glove with natural latex coating.
  • Palm coated with crinkle finish, fully coated thumb
  • Open back reduces perspiration build up
  • Good abrasion and cut resistance
  • Very good resistance to puncture
  • Excellent wet and dry grip
  • High flexibility and comfort
Typical applications include: General purpose, light car/vehicule maintenance, light material handling, project completion.
Certifications: EN 388 (3241); Cat. II