Imprimer
VWR®, Water Sampling Bottles, Square
Fournisseur: VWR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VWR®
VWRL080171EA 267 EUR
VWRL080171 VWRL080125S VWRL080168 VWRL080172W VWRL080152 VWRL080131
VWR®, Water Sampling Bottles, Square
Flacons Flacons d'échantillonnage de l'eau
Water sampling bottles, made of PET with PP screw caps, sterile dosed, sterile and non sterile.
  • PET food grade
  • Full traceability,batch number and individual bottle number on irradiated bottles
  • Fully recyclable
  • Cap / neck diameter 32 mm
Lightweight, durable bottles. Tamper evident screw caps. Choice of dosed and undosed sterile and non sterile. Square bottles for easy storage and handling.