Imprimer
Automatic Kjeldahl distillation unit, UDK 149
Fournisseur: VELP Scientifica
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
701-0225EA 8080 EUR
701-0225
Automatic Kjeldahl distillation unit, UDK 149
Dispositifs de distillation Systèmes de distillation Kjeldahl
The UDK 149 is a more flexible solution for laboratories performing Kjeldahl distillation. It is fully automatic and can be easily connected to a large choice of external titrators. The unit operates automatically, after setting water, boric acid and sodium hydroxide addition, distillation time and the steam generation output level between 10 and 100%. Different automatic titrator models can be connected for direct output of the final result. The high precision pumps ensure constant accurate dosing of reagents.
  • Automatic NaOH, H₂O and H₃BO₃ addition
  • Delay time (Devarda alloy analysis) and selectable distillation time
  • Automatic titration vessel washing
  • Distillation and titration residues removal; distillation in series
  • Language selection
  • Safety lever and sensors and reagent level warning to protect the user
All UDK series distillation units have an alkali resistant techno polymer housing and accept different kinds of test tubes: straight tubes (100, 250, 400 ml and 1 L) or Kjeldahl balloon (500 ml).
Certifications: Conformity with multiple standards protocols AOAC, ISO, EPA, DIN etc.
Informations sur les accessoires: Optional accessories such as test tubes, test tube spacer, test tube connection, connection cables and IQ/OQ/PQ manuals are available on request.
Informations de livraison: Each unit comes ready to use and is supplied with 250 ml test tube, 250 ml collecting flask, pincer for test tubes, touch pen and protective film for touch screen.