Imprimer

Eau, MINIVERSOL®, stérile

Marques : LABORATOIRE AGUETTANT
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5


600427
AGUE600427EA 58.6 EUR
AGUE600427
Eau, MINIVERSOL®, stérile
Eau

Sterile water is ideal for rinsing of non-sterile wounds, superficial burns, reusable medical devices and instruments.

Formule: H₂O
Poids moléculaire: 18,02 g/mol
Numéro MDL: MFCD00011332
Numéro CAS: 7732-18-5

Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR