Imprimer
Ultrasonic baths, Elmasonic X-tra TT series
Fournisseur: Elma
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Elmasonic
ELMA1076478EA 2820 EUR
ELMA1076478 ELMA1076475 ELMA1076477 ELMA1076476
Ultrasonic baths, Elmasonic X-tra TT series
Bains à ultrasons
Elmasonic xtra TT tabletop ultrasonic devices are designed for use in production environments, workshops and servicing. All functions and cleaning parameters like time and heating preselection and ultrasonic frequency are arranged user-friendly on the control panel on the front of the device where users can set and monitor them at a glance.
  • Permanently integrated Sweep-function for uniform distribution of the ultrasonic effect in the entire ultrasonic bath
  • Switchable Dynamic-function increases the peak ultrasonic performance
  • Individually settable limit temperature with LED warning indicator on reaching the temperature
  • Clearly arranged, control unit that is protected from spraying water
  • Safety shutdown and temperature-controlled auto-start function
With their ultrasonic performance of 37 kHz, Elmasonic xtra TT devices are rated for continuous heavy-duty operation. The tanks are made of special stainless steel, which makes them robust with a long service life.