Imprimer
Adapter, tri-clamp, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BOHLD901-02EA 49.6 EUR
BOHLD901-02
Adapter, tri-clamp, BOLA
This fitting made of PTFE can be used for the transition of Tri-Clamp as per DIN 32 676 to a GL threaded neck.
  • Universal chemical resistance, the flowing product is only exposed to PTFE
  • Plate diameter, 25,5 mm
This fitting can be used for connection of hard walled tubing (such as glass, stainless steel) or fluoroplastic tubes (such as PTFE, PFA and FEP) with laboratory screw joints to Tri-Clamp components as per DIN 32 676.