Imprimer
Safety funnels with ball valve, without lid
Fournisseur: SCAT Europe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
221-0232EA 177 EUR
221-0232
Safety funnels with ball valve, without lid
Entonnoirs Entonnoirs de sécurité
HDPE, white or HDPE, black, electrically conductive.
  • Sieve for trapping stirrers or larger contaminant particles
  • Splash protection for uniform discharge without splashes
  • Ball valve - the ball floats and automatically closes after filling
  • Freely rotatable cap
  • Safety lance - ensures clean outflow without contaminating the container opening
Models in black are made of electrically conductive HDPE and have a ground connection to prevent sparking hazards. With electrically conductive funnels (black), the lance facilitates a safe additional grounding of the contents.