Imprimer
Petrochemical standards, sulphur in isooctane
Fournisseur: LGC Standards
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PROOCON150-430-109EA 454 EUR
PROOCON150-430-109 PROOCON150-430-102 PROOCON150-430-020 PROOCON150-430-030 PROOCON150-430-010
Petrochemical standards, sulphur in isooctane
Étalons Étalons pour l'industrie pétrolière
These standards are designed for analysis of sulphur and chlorine in a wide variety of petroleum products by XRF, ICP and other analytical techniques.
All standards are prepareed on a weight/weight basis in high purity hydrocarbon oil with a 70 cSt viscosity at 40 °C (75 blank oil).