Imprimer
Buccal sample collection device, Whatman™ EasiCollect™+
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WHATWB120472EA 405 EUR
WHATWB120472
Buccal sample collection device, Whatman™ EasiCollect™+
EasiCollect™+ enables simple, reliable buccal sample collection and buccal DNA protection for DNA reference and other samples.
  • Updated EasiCollect™ design provides uniform buccal sample collection and transfer to FTA™ paper
  • Includes an Indicating FTA™ Card that changes colour from pink to white upon sample addition
  • Allows for direct STR amplification
  • Compatible with Hamilton Robotics’ easyPunch™ automation and Cytiva UniCore Punch for manual processing
  • Enables longterm archiving of DNA reference samples at room temperature with over 20 years of blood and 12 years of buccal real-time data support
EasiCollect™+ is an all-in-one single use product for the collection and storage of cheek cells. Cells are captured on the foam applicator by swabbing the inside of both cheeks and then transferred to an integral Indicating FTA™ Card. FTA™ Cards are treated with a proprietary chemical mix that breaks open cells and releases DNA. The same chemical mix then protects the DNA from damage. The Indicating FTA™ Cards also contains a dye that turns white when colourless liquids are applied to the card. This provides an easy method for sample location.
FTA™ chemistry lyses cells on contact, denatures proteins, and protects DNA from degradation. DNA remains tightly bound to the FTA™ Card while proteins and inhibitors are washed away. Once dried, the FTA™ Card can be removed from the EasiCollect™+ device and archived. The slim design of the card enables high-capacity storage.