Imprimer

Anti-APOA4 Mouse Monoclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI51-307EA 480 EUR
PRSI51-307
Anti-APOA4 Mouse Monoclonal Antibody
Anticorps
APOA4 antibody can be used in ELISA starting at 1:5000 - 1:25000, Western Blot starting at 1:2000 - 1:5000, immunohistochemistry starting at 10 - 20 μg/mL, and immunoprecipitation

0.03% sodium azide

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: APOA4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR