Imprimer

Anti-HDAC3 Mouse Monoclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI51-315EA 480 EUR
PRSI51-315
Anti-HDAC3 Mouse Monoclonal Antibody
Anticorps
HDAC3 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 μg/mL.

0.03% sodium azide

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HDAC3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR