Imprimer

Anti-MPO Mouse Monoclonal Antibody [clone: [9B12G7,4D8B12,9B12D9,9C11A5]]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI51-321EA 375 EUR
PRSI51-321
Anti-MPO Mouse Monoclonal Antibody [clone: [9B12G7,4D8B12,9B12D9,9C11A5]]
MPO antibody can be used in ELISA starting at 1:1000, and immunohistochemistry starting at 10 μg/mL.

0.03% sodium azide

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MPO
Clonality: Monoclonal
Clone: [9B12G7,4D8B12,9B12D9,9C11A5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR