Imprimer

Anti-MCAM Mouse Monoclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI51-346EA 480 EUR
PRSI51-346
Anti-MCAM Mouse Monoclonal Antibody
Anticorps
MCAM antibody can be used in immunofluorescence starting at 0.7 - 2 μg/mL, Western Blot starting at 1 - 3 μg/mL, and immunohistochemistry starting at 5 μg/mL.

Affinity Purified

PBS, pH 7.2, <=0.09% sodium azide, BSA and/or gelatin

Store MCAM antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles

Type:
Antigen: MCAM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR