Imprimer

Anti-CCR5 Mouse Monoclonal Antibody

Fournisseur: ProSci Inc.
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI51-348EA 619 EUR
PRSI51-348
Anti-CCR5 Mouse Monoclonal Antibody
Anticorps
CCR5 antibody can be used in Western Blot starting at 1 - 3 μg/mL, and immunohistochemistry starting at 20 μg/mL.

Affinity Purified

Aqeous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCR5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR