Imprimer

Anti-FCER1A Mouse Monoclonal Antibody

Fournisseur: ProSci Inc.
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI51-350EA 619 EUR
PRSI51-350
Anti-FCER1A Mouse Monoclonal Antibody
Anticorps
FCER1A antibody can be used in ELISA, Western Blot starting at 1 μg/mL, and immunohistochemistry starting at 5 μg/mL.

Affinity Purified

PBS, pH 7.2, 150 mM NaCl, 0.09% sodium azide

Store FCER1A antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles

Type:
Antigen: FCER1A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR