Imprimer

Anti-LMO2 Mouse Monoclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI51-354EA 480 EUR
PRSI51-354
Anti-LMO2 Mouse Monoclonal Antibody
Anticorps
LMO2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

Affinity Purified

Aqueous buffer, 0.09% sodium azide, contains stabilizer if necessary

Store LMO2 antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles

Type:
Antigen: LMO2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR