Imprimer

Anti-CD80 Mouse Monoclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI51-377EA 480 EUR
PRSI51-377
Anti-CD80 Mouse Monoclonal Antibody
Anticorps
CD80 antibody can be used in Western Blot, and immunohistochemistry starting at 10 μg/mL.

0.03% sodium azide

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CD80
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR