Imprimer

Anti-CDH2 Mouse Monoclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI51-407EA 0 EUR
PRSI51-407
Anti-CDH2 Mouse Monoclonal Antibody
Anticorps
N Cadherin antibody can be used in Western Blot at 0.5 - 2 μg/mL, immunohistochemistry starting at 20 μg/mL, and immunoprecipitation starting at 3 - 5 ng/1E6 cells.

0.03% sodium azide

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CDH2
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR