Imprimer
Whole blood, Parkinson’s disease (PD)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HCMSPB000F-2-PDEA 0 EUR
HCMSPB000F-2-PD
Whole blood, Parkinson’s disease (PD)
Whole blood is used for researching the causes, diagnosis, supportive care and treatment of Parkinson’s disease (PD). Collected from IRB-consented donors diagnosed with Parkinson’s disease (PD).
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Use immediately upon receipt.
Attention: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.