Imprimer
Whole blood, gastric cancer (GC)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HCMSPB000F-2-GCEA 0 EUR
HCMSPB000F-2-GC
Whole blood, gastric cancer (GC)
Whole blood used for researching the causes, diagnosis, supportive care and treatment of gastric cancer (GC). Patient samples can be used by researchers to investigate biomarker and diagnostic development, pre-clinical research validation, and drug toxicity and efficacy. The whole blood is collected from IRB-consented donors diagnosed with gastric cancer (GC).
No pre screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV Nucleic Acid, WNV Nucleic Acid, Trypanasoma cruzi and Syphilis.
Storage: Use immediately upon receipt.
Attention: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.