Imprimer
Hot air dryer
Fournisseur: Carl Roth
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ROTHY399.1EA 314 EUR
ROTHY399.1
Hot air dryer
Appareils chauffants Homogénéiseurs, broyeurs
This laboratory dryer with three heating and two cooling stages is for warm and cold air. Removal filter is present in the dryer for easy cleaning.
Informations de livraison: Incudes loop for hanging and attachable nozzle, 2 m power cable and power supply.