Imprimer
Ultrapure water system, Milli-Q® IQ 7000
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Milli-Q®
172-0029EA 0 EUR
MILLIQ7000 172-0029
Ultrapure water system, Milli-Q® IQ 7000
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
Milli-Q® IQ 7000 provides superior quality Type I ultrapure water from pretreated feed water with the combination of Jetpore® and IQnano® resin.
  • Simple and intuitive dispensing with the dispensing wheel
  • Environmentally-friendly mercury-free UV lamp oxidises organic contaminants
  • Superior ultrapure water consistently to the trace level
  • Easy data access: Users can see essential information (water quality, system status, warning, and more) on the Q-POD® unit screen at any time
  • Graphic display on the Milli-Q® production unit guides user through system operation, performance, trouble shooting and maintenance and can be password-protected
The system features two separate and distinct components; the production unit and the delivery point-of-use unit: The Milli-Q® production unit is a compact system that can be placed either on the bench, under the bench or on a wall. Maximum of four Q-POD® can be connected in series with the production unit.
Certifications: The Milli-Q® IQ 7000 system is delivered with a Certificate of Conformity ensuring that it has been built and tested fully assembled following Millipore Standard Operating Procedures and a Certificate of Calibration for the temperature and resistivity meters built in the system. Consumables are automatically delivered with a Certificate of Quality. Millipore’s manufacturing site is ISO 9001 v.2000 and ISO 140001 certified.
Informations de livraison: Order both the production unit and Q-POD® point-of-use dispenser for a complete system. Accessories and consumables, including wall mounting brackets, purification cartridges and replacement lamps, are sold separately.