Imprimer
Bi-metallic thermometer, H-B DURAC, SP Bel-Art
Fournisseur: Bel-Art Products, a part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Scienceware®
BELAB613104100EA 57.8 EUR
BELAB613104100
Bi-metallic thermometer, H-B DURAC, SP Bel-Art
Thermomètres Thermomètres analogiques
Thermometer features polycarbonate lens and beaker clip with built in adjustment tool in place of the probe cover with pocket clip.
  • Minimum immersion is 50 mm in liquids or 100 mm in gases
  • External adjustment nut allows for simple field calibration
  • Features accuracy to 1%, stainless steel case and stem
Informations de livraison: Includes a multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Test and Calibration.
Cdt: Individually packed.