Imprimer
Ductless filtering fume hoods, Captair® Smart and Midcap 832
Fournisseur: Erlab
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
135-1386EA 4000 EUR
ERLAB03NA0/11/BE ERLAB03NA0/11/AS
Ductless filtering fume hoods, Captair® Smart and Midcap 832
The new Captair® Smart ductless filtering hoods are designed with a simple and innovative way of communication. Smart Technology, simple and intuitive communication by light indicates the status of the hood. A simple light pattern will indicate if the airflow is compromised, if there is a fan failure or in case of a filter breakthrough.
  • Secured storage
  • Low power consumption
  • Real time sensors to detect main filter saturation with solvents, acids and formaldehye
Two configuration by double doors with shelves (120×1 l bottles) and pull-out doors with storage trays (100×1 l bottles).
The structure is made from corrosion resistant electro-galvanized steel coated with anti-acid polymer, sides and front panels are in chemical resistant acrylic
Types of carbon filters:
AS: For organics vapours
BE: Polyvalent for acid and organic vapours
Certifications: Meets AFNOR NF X 15-211:2009 standard : France EN 1822 : 1998 (HEPA H14 & ULPA U17 Filters).