Imprimer
Generic Pocket Colorimeter™ II with fixed wavelength
Fournisseur: Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HACH5870058EA 820 EUR
HACH5870058 HACH5870042 HACH5870065 HACH5870055 HACH5870045 HACH5870060 HACH5870050
Generic Pocket Colorimeter™ II with fixed wavelength
Rugged and reliable wavelength-specific filter photometer.
  • Simple: all functions are available via only four keys
  • Power: battery operation for a maximum of 2000 tests
  • Clear readings: even under difficult conditions, thanks to backlit display and large readout
  • Reliable results with no mains connection
  • Waterproof, lightweight, rugged
Intended for applications not included in standard analysis systems. The colorimeters are available in a range of wavelengths appropriate for many custom calibrations. In addition to the factory absorbance calibration (0 - 2,5 A), the instruments offer two user-entered, two-point, linear calibrations, and provide a choice of either 0,00 to 2,55 or 0,0 to 99,0 for the display value.
Pocket II mono-parameter colorimeters with ready to use reagents are also available for a broad range of parameters from A (aluminium) to Z (zinc). For details, please search ‘Pocket II’ and select ‘Colorimeters monoparameter, Pocket II’ to find the appropriate parameter in the table. Alternatively download the product literature from the web page.