Imprimer
Injector blade, No. 189
Fournisseur: Martor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MARO189.57EA 95.6 EUR
MARO189.57
Injector blade, No. 189
This blade is made from stainless steel.
  • Non corrosive
  • Two-sided ground-edge
  • Blade facets: Double bevel edge-ground
Cdt: 100 per magazine (5 magazines in multipack).